Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

401251

Sigma-Aldrich

IM-412

IM-412 primarily used in Inhibition.

Synonym(s):

IM-412, 3-(2-chlorobenzyl)-1,7-dimethyl-1H-imidazo[2,1-f]purine-2,4(3H,8H)-dione, TGF beta Inhibitor, IM-412

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClN5O2
Molecular Weight:
343.77
UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 3 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell permeable imidazoxanthine shown to decrease TGF-β stimulated reporter activity in a dose-dependent manner in HEK293 reporter cells, with an IC50 of 4.1 µM without cell cytotoxicity. Inhibits TGF-β receptor I (TβRI) and TGF-β receptor II (TβRII) expression and its downstream Smad2 and Smad3 phosphorylation as well as JNK and ERK activity. In addition, it elicits TGF-β-induced fibroblast differentiation with attenuated expression of several fibrotic markers such as α-SMA and fibronectin, and suppresses collagen accumulation in CCD-18Lu human normal lung fibroblasts.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Park, S., et al. 2010. Biochem. Biophy. Res. Comm.399, 268.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service