Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400062

Sigma-Aldrich

Anti-Glucose Transporter-3 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Glut-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

should not react with

rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A3(6515)

General description

Anti-Glucose Transporter-3, rabbit polyclonal, recognizes the ~45 kDa GLUT-3 protein. It is validated for use in ELISA, WB, IF, IP, and IHC (frozen sections).
Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~45 kDa Glut-3 protein.
Recognizes the ~45 kDa GLUT-3 protein.

Immunogen

a synthetic peptide corresponding to amino acids near the C-terminus of human GLUT-3, conjugated to KLH

Application

ELISA (0.5-1 µg/ml)

Immunoblotting (1-10 µg/ml)

Frozen Sections (2-20 µg/ml)

Immunofluorescence (2-20 µg/ml)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Does not cross-react with other Glut isoforms. This antibody has also been reported to work for immunoprecipitation. Variables associated with assay conditions will dictate the optimal working dilution.
Mueckler, M. 1994. Eur. J. Biochem. 219, 713.
Brant, A.M., et al. 1992. Biochem. Soc. Trans. 20, 236S.
Nagamatsu, S., et al. 1992. J. Biol. Chem. 267, 467.
Gould, G.W., et al. 1992. Diabetologia 35, 304.
Kayano, T., et al. 1988. J. Biol. Chem. 263, 15245.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mueckler
European journal of biochemistry, 219(3), 713-725 (1994-02-01)
Facilitative glucose transport is mediated by members of the Glut protein family that belong to a much larger superfamily of 12 transmembrane segment transporters. Six members of the Glut family have been described thus far. These proteins are expressed in
G W Gould et al.
Diabetologia, 35(4), 304-309 (1992-04-01)
Northern blot analysis of human tissues has demonstrated the expression of the brain-type glucose transporter isoform (GLUT 3) in liver, muscle and fat, raising the possibility that this transporter isoform may play a role in the regulation of glucose disposal
T Kayano et al.
The Journal of biological chemistry, 263(30), 15245-15248 (1988-10-25)
Complementary DNA clones encoding a glucose transporter-like protein have been isolated from a human fetal skeletal muscle cDNA library. The 496-amino acid fetal muscle glucose transporter-like protein has 64.4 and 51.6% identity with the previously described human erythrocyte/HepG2 and liver
Immunological identification of five members of the human facilitative glucose transporter family.
A M Brant et al.
Biochemical Society transactions, 20(3), 236S-236S (1992-08-01)
S Nagamatsu et al.
The Journal of biological chemistry, 267(1), 467-472 (1992-01-05)
Complementary DNAs encoding the mouse GLUT3/brain facilitative glucose transporter have been isolated and sequenced. The predicted amino acid sequence indicates that mouse GLUT3 is composed of 493 amino acids and has 83 and 89% identity and similarity, respectively, to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service