Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344550

Sigma-Aldrich

FTI-276

A highly potent and selective CAAX peptidomimetic of the carboxyl terminal of Ras proteins that inhibits farnesyl transferase (FTase) in vitro (IC₅₀ = 500 pM).

Synonym(s):

FTI-276, N-[2-phenyl-4-N[2(R)-amino-3-mercaptopropylamino benzoyl]-methionine, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N3O3S2
Molecular Weight:
433.59
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A highly potent and selective CAAX peptidomimetic of the carboxyl terminal of Ras proteins that inhibits farnesyl transferase (FTase) in vitro (IC50 = 500 pM). Inhibits geranylgeranyltransferase I (GGTase I) at much higher concentration (IC50 = 50 nM). Has been shown to selectively block the growth of a human lung carcinoma expressing oncogenic K-Ras in nude mice. Also inhibits oncogenic signaling and tumor growth of NIH 3T3 cells transformed with the Ras oncogene at 20 µM.

Biochem/physiol Actions

Cell permeable: no
Primary Target
FTase in vitro
Product does not compete with ATP.
Reversible: no
Target IC50: 500 pM against FTase in vitro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 1 week at -70°C.

Other Notes

Lantry, L.E., et al. 2000. Carcinogenesis21, 113.
Lerner, E.C., et al. 1995. J. Biol. Chem. 270, 26802.
Sun, J., et al. 1995. Cancer Res. 55, 4243.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service