Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329871

Sigma-Aldrich

Erythropoietin, Human

recombinant, expressed in CHO cells, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

Product Name

Erythropoietin, Human, Recombinant, CHO Cells,

description

Merck USA index - 14, 3688

Quality Level

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service