Recommended Products
antibody form
purified antibody
clone
polyclonal
purified by
affinity chromatography
species reactivity
rabbit, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
UniProt accession no.
General description
KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human Actin-related protein 2/3 complex subunit 2 (p34-ARC).
Specificity
Anti-Anti-p34-Arc\ARPC2 is a rabbit polyclonal antibody that detects Actin-related protein 2/3 complex subunit 2. It targets an epitope with in 14 amino acids from the C-terminal region.
Application
Epitope: C-terminus
Quality
Evaluated by Western Blotting in A431 cell lysate.
Western Blotting Analysis: 1 µg/mL of this antibody detected p34-Arc/ARPC2 in A431 cell lysate.
Western Blotting Analysis: 1 µg/mL of this antibody detected p34-Arc/ARPC2 in A431 cell lysate.
Physical properties
~34 kDa observed; 34.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Physical form
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Other Notes
Human. Predicted to react with Bovine, Mouse, and Rat based on 100% sequence homology.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service