Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860701P

Avanti

C17:0 GA2

powder

Synonym(s):

C17:0 Galabiosylceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H89NO13
CAS Number:
Molecular Weight:
876.21
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.28

Assay

99% (TLC)

form

powder

packaging

package of 1 × 500 μg (860701P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

Galabiosylceramide (Ga2cer) is a glycolipid and is produced in the luminal leaflet of the Golgi apparatus.

Biochem/physiol Actions

Galabiosylceramide (Ga2cer) is accumulated in larger amounts in Fabry′s disease.

Packaging

5 mL Clear Glass Sealed Ampule (860701P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topology of sphingolipid galactosyltransferases in ER and Golgi: trans bilayer movement of mono hexosyl sphingolipids is required for higher glycosphingolipid biosynthesis.
Burger KN, et al.
The Journal of Cell Biology, 133(1), 15-28 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service