Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860391P

Avanti

14:0-14:0-d27 PC

Avanti Research - A Croda Brand 860391P, powder

Synonym(s):

1-myristoyl-2-myristoyl-d27-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H45NO8PD27
CAS Number:
Molecular Weight:
705.10
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.12

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860391P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860391P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)COC(CCCCCCCCCCCCC)=O)=O

InChI key

CITHEXJVPOWHKC-QPUVRMJWSA-N

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Packaging

5 mL Amber Glass Screw Cap Vial (860391P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michel Roux et al.
Langmuir : the ACS journal of surfaces and colloids, 29(11), 3677-3687 (2013-02-16)
Amphiphilic cyclodextrins, with a cholesterol anchor (βChol) or an aspartic acid moiety esterified by two lauryl acyl chains (βDLC), were designed to combine the inclusion ability of the cyclodextrin cavity with the carrier properties of model membranes. Their insertion in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service