Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

857507P

Avanti

a15:0-i15:0 PG

Avanti Research - A Croda Brand

Synonym(s):

1-(12S-methylmyristoyl)-2-(13-methylmyristoyl)-sn-glycero-3-phospho-(1’-rac-glycerol) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H70NaO10P
CAS Number:
Molecular Weight:
716.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1  mg (857507P-1MG)
pkg of 1 × 5 mg (857507P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service