Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810342P

Avanti

22-NBD-Cholesterol

Avanti Research - A Croda Brand

Synonym(s):

22-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)amino)-23,24-bisnor-5-cholen-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38N4O4
CAS Number:
Molecular Weight:
494.63
MDL number:
UNSPSC Code:
12352211

biological source

synthetic

description

Technique(s): fluorescence

Assay

≥99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810342P-1MG)
pkg of 1 × 5 mg (810342P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

fluorescent lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@H](CNc1ccc([N+]([O-])=O)c2nonc12)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI key

DLWLXTLRGQWGPC-XIWKFBMMSA-N

Application

lipids research

Packaging

5 mL Amber Glass Screw Cap Vial (810342P-1MG);5 mL Amber Glass Screw Cap Vial (810342P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service