Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

700079P

Avanti

zymosterone

Avanti Research - A Croda Brand

Synonym(s):

cholesta-8(9),24-dien-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42O
CAS Number:
Molecular Weight:
382.62
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700079P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]1(CC2)C(CCC3=C1CC[C@@]4(C)C3CCC4[C@@H](CCC=C(C)C)C)CC2=O

InChI

1S/C27H42O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,19-20,23-24H,6,8-17H2,1-5H3/t19-,20?,23?,24?,26+,27-/m1/s1

InChI key

AUNLIRXIJAVBNM-ZSFXRWSJSA-N

Biochem/physiol Actions

Zymosterone is metabolized to cholesterol synthesis intermediate, zymosterol in the presence of enzyme hydroxysteroid (17β) dehydrogenase 7 (HSD17B7). Zymosterone conversion to zymosterol catalyzed by 3-keto sterol reductase (ERG27) is a key step in yeast ergosterol biosynthesis.

Packaging

5 mL Amber Glass Screw Cap Vial (700079P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service