Z742367
2MAG MIXcontrol MTP control device
AC/DC input 100-240 V, US 3-pin plug
Synonym(s):
control unit, power bank, power unit
About This Item
Recommended Products
material
stainless steel housing
stainless steel plate
feature
4 step power setting (25/50/75/100%)
packaging
pack of 1 ea
manufacturer/tradename
2mag 90250
parameter
0-40 °C temp. limit
100-2000 rpm speed
W × D × H
200 mm × 155 mm × 38 mm
plate size × W × L
× 200 mm × 120 mm, no of position= 12
weight
~1.4 kg
General description
Clear digital display for speed- and power settings, SoftStart for reliable centering and safe accelerating of the stirring bar.
Choose from one of the most recent versions:
Certificates of Analysis (COA)