Key Documents
Z278270
ACE Micro/Mini-Lab® pear-shaped flask
capacity 10 mL, single neck
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
pear-shaped flask
single-neck
Joint
joint ST/NS14/10 threaded
sterility
non-sterile
feature
closure type Holed cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9477-62
capacity
10 mL
General description
With cap, FETFE® O-ring, and septum.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Micro/Mini-Lab is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)