THS03146
ThalesNano MicroCatCart™ catalyst cartridge system
L × diam. 30 mm × 4 mm, 2.5%Pt, 2.5% Pd on carbon
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103817
NACRES:
NB.51
packaging
pkg of 6 ea
manufacturer/tradename
ThalesNano THS 03146
L × diam.
30 mm × 4 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
MicroCatCart is a trademark of ThalesNano Nanotechnology Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service