Key Documents
SYNF301200
Synthware single-neck round-bottom flask
200 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F301200, glass round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F301200
capacity
200 mL
200 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)