Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF57727705

Zirconium

foil, not light tested, 25x25mm, thickness 0.002mm, 99.2%

Synonym(s):

Zirconium, ZR000080

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Assay

99.2%

form

foil

manufacturer/tradename

Goodfellow 577-277-05

resistivity

40 μΩ-cm, 20°C

size × thickness

25x25 mm × 0.002 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daryoush Afzali et al.
Environmental monitoring and assessment, 186(6), 3523-3529 (2014-02-04)
In this study, a ternary Zr(IV) system with 2-(5-bromo-2-pyridylazo)-5-diethylaminophenol (5-Br-PADAP) and fluoride was chosen on the basis of dispersive liquid-liquid microextraction method. Zirconium was extracted into the fine droplets of dichlorobenzene as extracting solvent. These drops dispersed as a cloud
Muhammad Shahid et al.
Reviews of environmental contamination and toxicology, 221, 107-127 (2012-10-24)
Zirconium (Zr) is a transition metal that has both stable and radioactive isotopes.This metal has gained significant attention as a major pollutant of concern, partly because it has been prominent in the debate concerning the growing anthropogenic pressure on the
Oluwatayo F Ikotun et al.
Future medicinal chemistry, 3(5), 599-621 (2011-04-30)
Positron emission tomography, with its high sensitivity and resolution, is growing rapidly as an imaging technology for the diagnosis of many disease states. The success of this modality is reliant on instrumentation and the development of effective and novel targeted
Abnormal trace metals in man: zirconium.
H A Schroeder et al.
Journal of chronic diseases, 19(5), 573-586 (1966-05-01)
Joseph R Osborne et al.
The Journal of urology, 191(5), 1439-1445 (2013-10-19)
In this pilot study we explored the feasibility of (89)Zr labeled J591 monoclonal antibody positron emission tomography of localized prostate cancer. Before scheduled radical prostatectomy 11 patients were injected intravenously with (89)Zr-J591, followed 6 days later by whole body positron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service