GF35942632
Platinum
wire reel, 2m, diameter 0.15mm, annealed, may be used as one component of a thermocouple, 99.99%
Synonym(s):
Platinum, PT005119
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.99%
form
wire
manufacturer/tradename
Goodfellow 359-426-32
resistivity
10.6 μΩ-cm, 20°C
L × diam.
2 m × 0.15 mm
bp
3827 °C (lit.)
mp
1772 °C (lit.)
density
21.45 g/cm3 (lit.)
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jianbo Wu et al.
Accounts of chemical research, 46(8), 1848-1857 (2013-07-03)
An efficient oxygen reduction reaction (ORR) offers the potential for clean energy generation in low-temperature, proton-exchange membrane fuel cells running on hydrogen fuel and air. In the past several years, researchers have developed high-performance electrocatalysts for the ORR to address
Yanlong Yang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 2905-2921 (2013-12-18)
The relationship between the ERCC1/2 single nucleotide polymorphisms (SNPs) and the clinical outcomes of the platinum-based chemotherapy in the non-small cell lung cancer (NSCLC) is still inconsistent and inconclusive despite extensive investigations have been conducted to address this question. In
Yanqin Cao et al.
Journal of nanoscience and nanotechnology, 14(2), 1194-1208 (2014-04-23)
Porous platinum, that has outstanding catalytic and electrical properties and superior resistant characteristics to corrosion, has been widely applied in chemical, petrochemical, pharmaceutical, electronic, and automotive industries. As the catalytic activity and selectivity depend on the size, shape and structure
Johnson J Liu et al.
Current cancer drug targets, 12(8), 962-986 (2012-07-17)
Membrane transporters govern the movement of drugs and their metabolites across biological membranes, thereby determining their pharmacokinetics, efficacy and adverse drug reactions. Platinum-based anticancer drugs are a mainstay of chemotherapy for many human malignancies. However, their clinical utility is limited
Dalian Ding et al.
Anatomical record (Hoboken, N.J. : 2007), 295(11), 1851-1867 (2012-10-10)
Cisplatin, carboplatin, nedaplatin, and oxaliplatin are widely used in contemporary oncology; however, their ototoxic and neurotoxic side effects are quite different as discussed in this review. Cisplatin is considered the most ototoxic, but despite its reputation, the magnitude of hair
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service