Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF35916868

Palladium

foil, not light tested, 100x100mm, thickness 0.03mm, as rolled, 99.95%

Synonym(s):

Palladium, PD000261

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 359-168-68

resistivity

9.96 μΩ-cm, 20°C

size × thickness

100x100 mm × 0.03 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Doháňošová et al.
Molecules (Basel, Switzerland), 18(6), 6173-6192 (2013-05-28)
The palladium (II)-catalysed reactions of alkenols and aminoalkenols such as oxycarbonylations or bicyclisations are powerful methods for the construction of oxygen and nitrogen-containing heterocyclic compounds. This review highlights recent progress in the development of the asymmetric palladium(II)-catalysed Wacker-type cyclisations of
Simon De Corte et al.
Microbial biotechnology, 5(1), 5-17 (2011-05-11)
While precious metals are available to a very limited extent, there is an increasing demand to use them as catalyst. This is also true for palladium (Pd) catalysts and their sustainable recycling and production are required. Since Pd catalysts exist
M Angeles Fernández-Ibañez et al.
Molecules (Basel, Switzerland), 18(9), 10108-10121 (2013-08-27)
The dual activation of simple substrates by the combination of organocatalysis and palladium catalysis has been successfully applied in a variety of different asymmetric transformations. Thus, the asymmetric a-allylation of carbonyl compounds, a-fluorination of acyl derivatives, decarboxylative protonation of β-dicarbonyl
Masahiro Yoshida
Chemical & pharmaceutical bulletin, 60(3), 285-299 (2012-03-03)
It is known that propargylic compounds having an ester and a halide at the propargylic positions react with palladium complexes leading to π-propargylpalladium and allenylpalladium complexes, which cause various transformations in the presence of the reactants. The aim of the
Mélanie Platon et al.
Chemical Society reviews, 41(10), 3929-3968 (2012-03-27)
A survey highlighting the most recent palladium catalytic systems produced and their performances for progress in direct synthesis of indole backbones by heterocarbocyclization of reactive substrates is provided. The discussion is developed in relation with the principles of sustainable chemistry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service