Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF24896111

Copper

rod, 500mm, diameter 9.5mm, as drawn, 99.99+%

Synonym(s):

Copper, CU007955

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
12141711
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

rod

manufacturer/tradename

Goodfellow 248-961-11

resistivity

1.673 μΩ-cm, 20°C

L × diam.

500 mm × 9.5 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donita C Brady et al.
Nature, 509(7501), 492-496 (2014-04-11)
The BRAF kinase is mutated, typically Val 600→Glu (V600E), to induce an active oncogenic state in a large fraction of melanomas, thyroid cancers, hairy cell leukaemias and, to a smaller extent, a wide spectrum of other cancers. BRAF(V600E) phosphorylates and
Hiroki Serizawa et al.
Organic letters, 16(13), 3456-3459 (2014-06-14)
The direct synthesis of pentafluoroethyl copper (CuC2F5) from a cuprate reagent and ethyl pentafluoropropionate as one of the most economical and useful pentafluoroethyl sources was accomplished. The advantages of this method are; all the reagents employed are low-cost and operationally
Gwilherm Evano et al.
Natural product reports, 30(12), 1467-1489 (2013-10-25)
Copper-catalyzed Ullmann-Goldberg-type cross-coupling reactions have undergone nothing short of a renaissance over the last decade and an impressive number of procedures are now available for the formation of C-N, C-O and C-S bonds with remarkable efficiencies and surgical precision. These
Nathalie Adam et al.
Environmental pollution (Barking, Essex : 1987), 194, 130-137 (2014-08-12)
In this study the uptake of ZnO and CuO nanoparticles by Daphnia magna was tested. Daphnids were exposed during 48 h to acute concentrations of the nanoparticles and corresponding metal salts. The Daphnia zinc and copper concentration was measured and the
Marta Ugarte et al.
Survey of ophthalmology, 58(6), 585-609 (2013-10-29)
The essential trace metals iron, zinc, and copper play important roles both in retinal physiology and disease. They are involved in various retinal functions such as phototransduction, the visual cycle, and the process of neurotransmission, being tightly bound to proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service