Key Documents
DWK209630
Wheaton® Leak Resistant Bottle
capacity 1000 mL, high-density polyethylene bottle, amber bottle, wide-mouth bottle, bottle diam. × H 91 mm × 199 mm, 63-415
Synonym(s):
209630
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
amber bottle
high-density polyethylene bottle
wide-mouth bottle
packaging
pkg of 48 ea
manufacturer/tradename
WHEATON® 209630
bottle diam. × H
91 mm × 199 mm
capacity
1000 mL
Features and Benefits
Superior double seal closure for leak resistance
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)