Key Documents
DWK209546
Wheaton® Leak Resistant Bottle
capacity 60 mL, high-density polyethylene bottle, natural bottle, wide-mouth bottle, bottle diam. × H 39 mm × 88 mm, 28-410
Synonym(s):
209546
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
high-density polyethylene bottle
natural bottle
wide-mouth bottle
packaging
pkg of 72 ea
manufacturer/tradename
WHEATON® 209546
bottle diam. × H
39 mm × 88 mm
capacity
60 mL
Features and Benefits
Superior double seal closure for leak resistance
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)