D19303
4,5-Diamino-6-hydroxypyrimidine hemisulfate salt
Synonym(s):
5,6-Diamino-4-pyrimidinone hemisulfate salt, 5,6-Diamino-4-pyrimidinol hemisulfate salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H12N8O2 · H2SO4
CAS Number:
Molecular Weight:
350.31
Beilstein:
3771022
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
crystals
Quality Level
mp
270 °C (dec.) (lit.)
SMILES string
OS(O)(=O)=O.Nc1ncnc(O)c1N.Nc2ncnc(O)c2N
InChI
1S/2C4H6N4O.H2O4S/c2*5-2-3(6)7-1-8-4(2)9;1-5(2,3)4/h2*1H,5H2,(H3,6,7,8,9);(H2,1,2,3,4)
InChI key
SVJZVGHPJPXEJQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service