Key Documents
CLS7715
Corning® thermometer stopper
rubber
Synonym(s):
Corning stopper
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
material
rubber
manufacturer/tradename
Corning 7715
O.D. × I.D.
17 mm × 13 mm
General description
This Corning rubber thermometer holder has a 17 mm O.D. and a 13 mm I.D. and is designed for use with the Corning CLS7711 mercury thermometer.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)