Key Documents
BR705448
BRAND® Transferpette® multichannel, electronic pipette
new
12-channel, volume 50—1250 μL, AC/DC input 100 V / 240 V AC, 50—60 Hz, universal plug set
Synonym(s):
8-channel, Transferpette, electronic, micropipette, micropipettor, multichannel, pipet, pipette, pipettor
About This Item
UNSPSC Code:
41121503
Sterilization Compatibility
autoclavable compatible
feature
12-channel
packaging
pack of 1 ea
manufacturer/tradename
BRAND 705448
volume
50—1250 μL
battery power
(Nickel metal hydride 3 cells)
General description
Transferpette® electronic pipettes combine the globally proven properties of mechanical piston-operated air displacement pipettes by BRAND with the advantages of an electronic device.
Features and Benefits
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
  • Easy calibration technique for making quick adjustments to the Transferpette® electronic, without tools.
  • Ergonomic, handy design
  • Clear and intuitive operation
  • Up to 4k pipetting cycles on a single charge
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)