Key Documents
BR704548
BRAND® replacement pump assembly for seripettor® pro
size 10 mL
Synonym(s):
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
UNSPSC Code:
40151500
NACRES:
NB.01
material
(Polyphenylene Oxide/polyetherimide (UV protection))
(Hastalloy (stainless))
packaging
pack of 1 ea
manufacturer/tradename
BRAND 704548
size
10 mL
General description
The BRAND® Replacement Pump Assembly for the Seripettor® Pro is designed for precise and efficient liquid handling in laboratory applications. This actuating unit is constructed from high-quality materials, including polyphenylene oxide (PPO) and polyetherimide (PEI) for UV protection, ensuring durability and longevity. The assembly features a Hastelloy® lifting spring, which enhances performance and reliability. This replacement pump assembly is essential for maintaining the functionality of your Seripettor® Pro, making it an indispensable tool for researchers and technicians seeking accurate and efficient liquid dispensing.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Hastelloy is a registered trademark of Haynes International, Inc.
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)