Key Documents
BAF126280000
Pouring spout with fingertip control
pack of 12 ea, high-density polyethylene
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
material
high-density polyethylene
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F12628-0000
General description
Chemical resistant, high-density polyethylene spout has a small air hole that allows finger tip control of pouring speed.
  • Fits 38 mm tall buttress closure bottles
Choose from one of the most recent versions:
Certificates of Analysis (COA)