Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

940682

Sigma-Aldrich

TissueFab® tyramine functionalized poly(amino acid)

hydrogel and bioink precursor, for tissue engineering

Synonym(s):

Amino acid, Amino acid polymer, Aspartic acid, Bioink precursor polymers, Hydrogel materials, Poly(aspartic acid), Tyramine functionalized polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H18N2O5)m(C12H14N2O3)n
UNSPSC Code:
12352200

Quality Level

form

powder, crystals or chunks

color

white to off-white

application(s)

3D bioprinting
(Tissue engineering, 3D Cell culture)

compatibility

(cell culture)

storage temp.

−20°C

General description

Rethink your bioinks with TissueFab®About the material: These TissueFab® tyramine functionalized poly(amino acid) is a xeno-free (animal origin free), biodegradable and biocompatible polymer that been developed and tested for hydrogel synthesis for tissue engineering and other biomedical applications.


Printing and crosslinking parameters:

  • Compatible for cell encapsulation and hydrogel formation
  • Crosslinked via a ruthenium photoinitiatior system
The TissueFab® tyramine functionalized poly(amino acid) is characterized by its biocompatibility and biodegradability, making it well-suited for a broad spectrum of applications in the biomedical field. This poly(amino acid) is synthesized through the ring opening of polysuccinimide using a nucleophilic reagent. The presence of amide linkages in these polymers results in peptide-like structures that are prone to cleavage and degradation in the body, leading to the formation of amino acids or small peptides. The tyramine functional group within the polymer provides sites for further chemical modification or conjugation with bioactive molecules, such as drugs, peptides, and targeting ligands. For example, the free amine group on tyramine can undergo acylation with acid chlorides or anhydrides to form amide derivatives. Additionally, tyramine can participate in enzymatic or chemical crosslinking reactions to form covalent bonds between tyramine molecules or with other biomolecules. Furthermore, this tyramine functionalized poly(amino acid) can be photo-oxidized by the Ruthenium photoinitiator kit (Cat#916881) into reactive intermediates, which can then crosslink with proteins or adjacent tyramine to form a cell compatible hydrogel.
More information and a step by step protocol can be found under the "More Documents" section of this page.

Application

The TissueFab® tyramine functionalized poly(amino acid) xeno-free polymer is:
  • Biocompatible and biodegradable precursor for hydrogel and bioink development
  • Suitable for a wide range of biomedical applications, including 3D cell culture, tissue engineering, in vitro drug screening, and regenerative medicine research
  • Enables the development of xeno-free polymers for applications in advanced cell culture and tissue engineering
  • Ideal for creating biocompatible materials that support cell growth and tissue regeneration in a xenogeneic-free environment

Features and Benefits

This TissueFab® hydrogel and bioink precursor has been developed and tested to meet strict criteria to best suit researchers′ needs. This includes:
  • Xeno-free, animal-origin free
  • Tunable
  • Cell viability
  • Biocompatibility and biodegradability
  • Mechanical and rheological properties
  • Crosslinkability

Legal Information

TISSUEFAB is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service