940283
TissueFab® collagen bioink kit
neutral, Viscofan Fibercoll-Flex® N
Synonym(s):
Insoluble collagen bioink
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
form
(viscous suspension)
composition
from bovine dermis
application(s)
(3d bioprinting)
(tissue engineering)
compatibility
cell culture
extrusion-based 3D bioprinting
storage temp.
2-8°C
Related Categories
General description
Rethink your bioinks with TissueFab®
About the material: Fibercoll-Flex-N(R) is a bioink formulated of pure, fibrillary collagen type I, making it ideal for printing highly biocompatible scaffolds with embedded cells.
Printing and crosslinking parameters:
About the material: Fibercoll-Flex-N(R) is a bioink formulated of pure, fibrillary collagen type I, making it ideal for printing highly biocompatible scaffolds with embedded cells.
Printing and crosslinking parameters:
- Made of acidic collagen fibers, the Fibercoll-Flex-N(R) is neutralized before printing, enabling the generation of bioprints with encapsulated cells at physiological conditions without a methacrylation curing step
- Easy and efficient extrusion-based printing at room temperature or up to 37°C, no cooling devices for printer required
Application
The Fibercoll-Flex-N® can be used for any bioprinting applications including:
- 3D cell culture in scaffolds of customized shape and stiffness
- Complex tissue models
- Tumor models
- Tissue engineering and regenerative medicine
- Cell based assays such as drug metabolism for drug discovery or drug screening applications
Features and Benefits
Key Features and Benefits: Each TissueFab(R) material is developed and tested to meet strict criteria to best suit researchers′ needs. This includes:
- Printability and fidelity
- Cell viability
- Biocompatibility
- Sterility
- Mechanical and rheological properties
Legal Information
TISSUEFAB is a registered trademark of Merck KGaA, Darmstadt, Germany
VISCOFAN FIBERCOLL-FLEX is a registered trademark of VISCOFAN, S.A.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service