Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

938831

Sigma-Aldrich

NanoFabTx Polymers for Microneedles

GelMA hydrogel kit

Synonym(s):

Gelatin methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Application

NanoFabTx Polymers for Microneedles are carefully selected polymers that can be used to formulate microneedle patches. Microneedles are tiny projections of polymeric needles that can vary in length, pitch, and array for topical drug delivery applications. They are typically composed of biodegradable synthetic or natural polymers at a variety of suitable concentrations.
Gelatin methacrylate (GelMA) is one of the most common natural polymers used in a variety of biomedical applications due to its suitable biological properties and tunable physical characteristics. GelMA can be used to form hydrogels via photopolymerization with photoiniatiors such as lithium phenyl-2,4,6-trimethylbenzoylphosphinate (LAP). GelMA hydrogel microneedles have been used to encapsulate and delivery a variety of therapeutics including small molecules, exosomes, and proteins.

Features and Benefits

The NanoFabTx Polymers for Microneedles can be used with microneedle templates to formulate microneedles for drug delivery applications. The selected polymer will enable users to:


1)save time in selecting the polymer and provide a starting point to formulate microneedles by avoiding lengthy trial-and-error optimization.
2)formulate microneedles for various drug delivery applications.

Preparation Note

For more information on the protocols, please refer to Product Information under the document section of this page.

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service