Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

937355

Sigma-Aldrich

ImPy-Trip-IPr*

greener alternative

≥95%

Synonym(s):

2‐[2,6‐Bis(diphenylmethyl)‐4‐methylphenyl]‐5‐[2,4,6‐tris(propan‐2‐yl)phenyl]‐2H‐4Λ5‐imidazo[1,5‐a]pyridin‐4‐ylium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H55ClN2
CAS Number:
Molecular Weight:
779.49
UNSPSC Code:
12352142

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Cross Couplings

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

functional group

(Carbene)

greener alternative category

SMILES string

CC(C=C1C(C2=CC=CC=C2)C3=CC=CC=C3)=CC(C(C4=CC=CC=C4)C5=CC=CC=C5)=C1N6C=[N+]7C(C=CC=C7C8=C(C(C)C)C=C(C(C)C)C=C8C(C)C)=C6.[Cl-]

InChI

1S/C55H55N2.ClH/c1-37(2)45-33-47(38(3)4)54(48(34-45)39(5)6)51-30-20-29-46-35-56(36-57(46)51)55-49(52(41-21-12-8-13-22-41)42-23-14-9-15-24-42)31-40(7)32-50(55)53(43-25-16-10-17-26-43)44-27-18-11-19-28-44;/h8-39,52-53H,1-7H3;1H/q+1;/p-1

InChI key

RFDIMPRGVRMUES-UHFFFAOYSA-M

General description

ImPy-Trip-IPr* is a member of a class of biaryl NHC ligands utilizing sterics to facilitate a well-defined L-shaped NHC. The unique shape allows for the development of cross-coupling reactions not possible with other NHC ligands, as well as an improvement in catalytic efficiency with many existing methods. Readily forms complexes with Pd, as well as Au, Cu, Rh, and other metals.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Active in the following reactions:
  • Suzuki-Miyarua cross-coupling
  • Buchwald-Hartwig amination
  • Hydroxylation of aryl chlorides

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service