Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

936030

Sigma-Aldrich

Sodium triflate

new
greener alternative

≥99.5% trace metals basis, anhydrous, battery grade

Synonym(s):

Sodium trifluoromethanesulfonate, NaTfO, Sodium triflate, Trifluoromethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Na
CAS Number:
Molecular Weight:
172.06
Beilstein:
3728797
MDL number:
UNSPSC Code:
12352104

grade

battery grade

Quality Level

Assay

≥99.5% trace metals basis

form

powder or crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤5000 ppm trace metals

mp

253-255  °C
253-255 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Na+].[O-]S(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S.Na/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1

InChI key

XGPOMXSYOKFBHS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium trifluoromethanesulfonate, or sodium triflate (NaTfO), is a white, crystalline salt. It is hygroscopic and deliquescent. NaTfO is soluble in water, acetonitrile, some carbonates (including dimethyl carbonate and propylene carbonate, but insoluble in diethyl carbonate), glymes (including monoglyme, diglyme, and triglyme), dimethoxyethane, and most polar organic solvents. It is thermally stable and melts at 253 - 255 °C. NaTfO can be prepared by reaction of sodium sulfate and barium trifluoromethanesulfonate. Alternatively, it can be prepared by reaction between sodium dithionite and bromotrifluoromethane.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Our sodium trifluoromethanesulfonate offers the high purity and low water content ideal for use as an electrolytic salt in sodium-ion batteries. NaTfO is compatible with many cathode and anode materials used in sodium-ion batteries, including sodium metal anodes used in sodium metal batteries Additionally, NaTfO dissolved in diglyme has enabled the use of graphite in reversible sodium-ion batteries When electrolytes comprised of NaTfO dissolved in diglyme, tetraglyme, or 1,2-dimethoxyethane are used with graphite anodes, solvated sodium co-intercalates along with its anion facilitating fast kinetics and high charge rates Researchers have also found that sodium trifluoromethanesulfonate performs well with layered chalcogenides like FeS2 and CoSe2 when other electrolytes perform poorly. Lastly, high-purity NaTfO dissolved in diglyme and 1,3-dioxolane has shown promise as an electrolyte in sodium-oxygen batteries form a thin stable passivation film on the alkali metal anode.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service