Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

927732

Sigma-Aldrich

C5 Lenalidomide-difluoroPEG1-C4-piperazine Hydrochloride

≥95%

Synonym(s):

2-(2,2-Difluoro-3-(4-(piperazin-1-yl)butoxy)propoxy)-N-(2-(2,6-dioxopiperidin-3-yl)-1-oxoisoindolin-5-yl)acetamide Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35F2N5O6 · xHCl
Molecular Weight:
551.58 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

ligand

C5 Lenalidomide

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

O=C1N(C2CCC(NC2=O)=O)CC3=CC(NC(COCC(F)(F)COCCCCN4CCNCC4)=O)=CC=C31.Cl

InChI

1S/C26H35F2N5O6.ClH/c27-26(28,16-38-12-2-1-9-32-10-7-29-8-11-32)17-39-15-23(35)30-19-3-4-20-18(13-19)14-33(25(20)37)21-5-6-22(34)31-24(21)36;/h3-4,13,21,29H,1-2,5-12,14-17H2,(H,30,35)(H,31,34,36);1H

InChI key

NWCOGWMYGNQKPZ-UHFFFAOYSA-N

Application

Protein degrader building block C5 Lenalidomide-difluoroPEG1-C4-piperazine Hydrochloride enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a Cereblon (CRBN)-recruiting ligand, a fluorinated linker with both hydrophobic and hydrophilic moieties, and a pendant amine for reactivity with a carboxylic acid on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant amine, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.


Targeted Protein Degradation

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel P Bondeson et al.
Annual review of pharmacology and toxicology, 57, 107-123 (2016-10-13)
Protein homeostasis networks are highly regulated systems responsible for maintaining the health and productivity of cells. Whereas therapeutics have been developed to disrupt protein homeostasis, more recently identified techniques have been used to repurpose homeostatic networks to effect degradation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service