924938
25-Hydroxyvitamin D3-(23,24,25,26,27-13C5)
≥99 atom % 13C, ≥95% (CP)
About This Item
Recommended Products
isotopic purity
≥99 atom % 13C
Quality Level
Assay
≥95% (CP)
mass shift
M+5
SMILES string
C[C@@]12[C@](CC[C@]2([H])[C@H](C)C[13CH2][13CH2][13C](O)([13CH3])[13CH3])([H])/C(CCC1)=C/C=C(C[C@@H](O)CC3)\C3=C
InChI
1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1/i3+1,4+1,6+1,16+1,26+1
InChI key
JWUBBDSIWDLEOM-PQZGROPVSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service