Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900851

Lactose ureide-15N2

≥98 atom % 15N, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H2415N2O11
Molecular Weight:
386.32
UNSPSC Code:
12352201

isotopic purity

≥98 atom % 15N

Assay

≥98% (CP)

form

solid

mass shift

M+2

SMILES string

[15NH2]C([15NH][C@H](O[C@@H]1CO)[C@H](O)[C@@H](O)[C@@H]1O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)[C@@H](CO)O2)=O

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service