Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755486

Sigma-Aldrich

(-)-2-(2-Naphthylmethoxy)isopinocampheol

97%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

293-298 °C

functional group

ether
hydroxyl

storage temp.

2-8°C

InChI

1S/C21H26O2/c1-20(2)17-11-18(20)21(3,19(22)12-17)23-13-14-8-9-15-6-4-5-7-16(15)10-14/h4-10,17-19,22H,11-13H2,1-3H3/t17-,18-,19+,21-/m0/s1

InChI key

NPEWTQVRFLMGHP-HUUJSLGLSA-N

Application

Superior reagent for chiral aldol reactions

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service