71494
Sodium diformylamide
≥97.0% (NT)
Synonym(s):
Diformylimide sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HCO)2NNa
CAS Number:
Molecular Weight:
95.03
Beilstein:
3567185
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥97.0% (NT)
form
powder
functional group
amine
SMILES string
[Na+].O=C[N-]C=O
InChI
1S/C2H3NO2.Na/c4-1-3-2-5;/h1-2H,(H,3,4,5);/q;+1/p-1
InChI key
QJXDSDLNUKLDBP-UHFFFAOYSA-M
Application
Reactant for preparation of:
- Functionalized condensed benzazoles
- α-ketoheterocycles as inhibitors of Leishmania mexicana cysteine protease CPB
- INOS inhibitor PHA-399733 via Bucherer-Bergs hydantoin synthesis
- Antibiotic resistance (C β-lactamase AmpC ) inhibitor analogues, with improved cell permeability properties
- BINOL- and H8-BINOL-based aryl phosphites as ligands in palladium-catalyzed asymmetric allylation and amination reactions
- Precursors of αVβ3 integrin antagonists
Other Notes
Modified Gabriel reagent for the synthesis of amines; mild hydrolysis to release the amine
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H. Yinglin et al.
Synthesis, 122-122 (1990)
B. Gohrmann et al.
J. Prakt. Chem., 331, 819-819 (1989)
Synthesis, 615-615 (1990)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service