Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

683566

Sigma-Aldrich

1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester

97%

Synonym(s):

Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21BN2O4
CAS Number:
Molecular Weight:
280.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.4814

functional group

ester

SMILES string

CCOC(=O)Cn1cc(cn1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C13H21BN2O4/c1-6-18-11(17)9-16-8-10(7-15-16)14-19-12(2,3)13(4,5)20-14/h7-8H,6,9H2,1-5H3

InChI key

YUEZJHOSHBTWPV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service