586188
Boc-Asp-OH-3-13C
99 atom % 13C
Synonym(s):
N-(tert-Butoxycarbonyl)-L-aspartic acid-3-13C, L-Aspartic acid-3-13C, N-t-Boc derivative
About This Item
Recommended Products
isotopic purity
99 atom % 13C
form
solid
optical activity
[α]20/D -7.3°, c = 2 in DMF
technique(s)
bio NMR: suitable
mp
116-118 °C (lit.)
application(s)
peptide synthesis
functional group
Boc
mass shift
M+1
SMILES string
CC(C)(C)OC(=O)N[C@@H]([13CH2]C(O)=O)C(O)=O
InChI
1S/C9H15NO6/c1-9(2,3)16-8(15)10-5(7(13)14)4-6(11)12/h5H,4H2,1-3H3,(H,10,15)(H,11,12)(H,13,14)/t5-/m0/s1/i4+1
InChI key
KAJBMCZQVSQJDE-RGUYRYBTSA-N
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service