About This Item
Recommended Products
isotopic purity
98 atom % D
Quality Level
mass shift
M+3
SMILES string
[K+].[2H][B-]([2H])([2H])C#N
InChI
1S/CH3BN.K/c2-1-3;/h2H3;/q-1;+1/i2D3;
InChI key
ZNBXOJRKRYPEJV-MUTAZJQDSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service