Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

296015

Sigma-Aldrich

Acetonitrile-2-13C, d3

99 atom % 13C, 99.5 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD3CN
CAS Number:
Molecular Weight:
45.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
99.5 atom % D

Quality Level

Assay

99% (CP)

bp

81 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

mass shift

M+4

SMILES string

[2H][13C]([2H])([2H])C#N

InChI

1S/C2H3N/c1-2-3/h1H3/i1+1D3

InChI key

WEVYAHXRMPXWCK-KQORAOOSSA-N

Packaging

Packaged in prescored ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service