Merck
  • In-vitro evidence of enhanced breast cancer resistance protein-mediated intestinal urate secretion by uremic toxins in Caco-2 cells.
In-vitro evidence of enhanced breast cancer resistance protein-mediated intestinal urate secretion by uremic toxins in Caco-2 cells.
The Journal of pharmacy and pharmacology (2014-12-18)
Yang Lu, Takeo Nakanishi, Atsushi Hosomi, Hisakazu Komori, Ikumi Tamai
ABSTRACT
It has been reported that intestinal urate excretion is increased at chronic kidney disease (CKD) state. In this report, whether uremic toxins are involved in the upregulation of intestinal breast cancer resistance protein (BCRP), an intestinal urate exporter, was examined. Uremic toxins that were increased at least 15-fold at CKD state were selected for investigation. Caco-2 cells were exposed to these uremic toxins at clinically relevant concentrations. mRNA was quantified by real-time PCR, and flow cytometry was utilized to measure BCRP protein and function in Caco-2 cells. Transcellular secretory transport of [(14) C]urate was determined utilizing Transwell studies after uremic toxin exposure. Indoxyl sulfate (IS) treatment alone resulted in ∼ 3-fold increase in BCRP mRNA in Caco-2 cells. Membrane protein expression of BCRP in Caco-2 cells also was increased by 1.8-fold after treatment with IS. Intracellular accumulation of pheophorbide A, a selective BCRP substrate, was decreased by 22% after IS treatment for 3 days. Consistent with these findings, transcellular secretory transport of urate across Caco-2 cell monolayers was increased by 22%. Intestinal urate secretion may be increased at CKD state partially by upregulation of intestinal BCRP by uremic toxins such as IS.
Product Description
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylguanidine hydrochloride, 98%
Sigma-Aldrich
CH-223191
Sigma-Aldrich
p-Cresol, ≥99%, FG
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
p-Cresol, 99%
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland