Skip to Content
Merck
All Photos(1)

Key Documents

1138493

USP

Clocortolone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9-Chloro-6α-fluoro-11β,21-dihydroxy-16α-methyl-1,4-pregnadien-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28ClFO4
CAS Number:
Molecular Weight:
410.91
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

clocortolone

form

solid

manufacturer/tradename

USP

SMILES string

F[C@H]1C[C@@H]2[C@@]([C@@]4(C1=CC(=O)C=C4)C)([C@H](C[C@]3([C@H]2C[C@H]([C@@H]3C(=O)CO)C)C)O)Cl

InChI

1S/C22H28ClFO4/c1-11-6-13-14-8-16(24)15-7-12(26)4-5-21(15,3)22(14,23)18(28)9-20(13,2)19(11)17(27)10-25/h4-5,7,11,13-14,16,18-19,25,28H,6,8-10H2,1-3H3/t11-,13+,14+,16+,18+,19-,20+,21+,22+/m1/s1

InChI key

YMTMADLUXIRMGX-RFPWEZLHSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service