Skip to Content
Merck
All Photos(1)

Key Documents

PHL84191

Protocetraric acid

phyproof® Reference Substance

Synonym(s):

4-Formyl-3,8-dihydroxy-9-(hydroxymethyl)-1,6-dimethyl-11-oxo-11H-dibenzo[b,e][1,4]dioxepin-7-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14O9
CAS Number:
Molecular Weight:
374.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

SMILES string

O1c2c(c(c(c(c2C)C(=O)O)O)CO)OC(=O)c3c1c(c(cc3C)O)C=O

InChI

1S/C18H14O9/c1-6-3-10(21)8(4-19)15-11(6)18(25)27-16-9(5-20)13(22)12(17(23)24)7(2)14(16)26-15/h3-4,20-22H,5H2,1-2H3,(H,23,24)

InChI key

VOXMONAUSQZPTP-UHFFFAOYSA-N

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service