Skip to Content
Merck
All Photos(1)

Key Documents

95713

Supelco

1,4-Butanediol dimethacrylate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO2CH2CH2-]2
CAS Number:
Molecular Weight:
226.27
MDL number:

grade

analytical standard

Quality Level

Assay

≥94.5% (GC)

technique(s)

HPLC: suitable

refractive index

n20/D 1.456 (lit.)

bp

132-134 °C/4 mmHg (lit.)

density

1.023 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCCCOC(=O)C(C)=C

InChI

1S/C12H18O4/c1-9(2)11(13)15-7-5-6-8-16-12(14)10(3)4/h1,3,5-8H2,2,4H3

InChI key

XOJWAAUYNWGQAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Components

contains approx. 200 ppm MEHQ as inhibitor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service