Skip to Content
Merck
All Photos(1)

Key Documents

78347

Supelco

Creatine

analytical standard

Synonym(s):

(α-Methylguanido)acetic acid, N-Amidinosarcosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)N(CH3)CH2CO2H
CAS Number:
Molecular Weight:
131.13
Beilstein:
907175
MDL number:

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages
personal care
pharmaceutical

format

neat

SMILES string

CN(CC(O)=O)C(N)=N

InChI

1S/C4H9N3O2/c1-7(4(5)6)2-3(8)9/h2H2,1H3,(H3,5,6)(H,8,9)

InChI key

CVSVTCORWBXHQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Creatine is a nitrogenous compound that acts as a high-energy reservoir for the rapid regeneration of ATP.
Creatine is a nitrogenous compound that acts as a high-energy reservoir for the rapid regeneration of ATP. Approximately 95% of creatine is found in skeletal muscle, primarily as phosphocreatine. Creatine can be acquired through dietary consumption or formed from L-arginine, glycine, and L-methionine in a multi-step reaction that occurs in the kidneys and liver. Creatine is then transported to muscle tissue. Creatine supplementation is used for the enhancement of sports performance, primarily by increasing muscle mass. Creatine is also being investigated as a treatment of neuromuscular diseases, where it may aid in neuroprotection and by improving the cellular bioenergetic state.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service