Key Documents
57472
Rheodyne® Port Adapter
About This Item
UNSPSC Code:
41115854
General description
Use the Port Adapter for connecting 1/16 in. tubing to a 1/8 in. valve port.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)