27413
Screw cap (open-top), polypropylene
yellow polypropylene hole cap, thread for 10-425, for use with 2 mL large opening vial (6.0 mm), pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
yellow polypropylene hole cap
feature
autoclavable
closure type black polypropylene hole cap
packaging
pkg of 100 ea
parameter
135 °C temp. limit
fitting
thread for 10-425
compatibility
for use with 2 mL large opening vial (6.0 mm)
Looking for similar products? Visit Product Comparison Guide
General description
These hole caps do not come with septa.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service