General description
We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency".
Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.
Specificity
Clone 2B15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Neural cell adhesion molecule L1 (L1CAM/CD171). It targets an epitope within 21 amino acids from the extracellular domain.
Immunogen
KLH-conjugated linear peptide corresponding to 21 amino acids from the extracellular domain of human Neural cell adhesion molecule L1 (L1CAM/CD171).
Application
Quality Control Testing
Evaluated by Western Blotting in MCF-7 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected L1CAM/CD171 in MCF-7 cell lysate.
Tested Applications
Affinity Binding Assay: A representative lot of this antibody bound L1CAM-CD171 peptide with a KD of 5.2 x 10-8 in an affinity binding assay.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected L1CAM/CD171 in MCF-7 cells.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Neural cell adhesion molecule L1 (UniProt: P32004; also known as N-CAM-L1, NCAM-L1, CD171) is encoded by the L1CAM (also known as CAML1, MIC5) gene (Gene ID: 3897) in human. L1CAM is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 20-1120), a transmembrane domain (aa 1121-1143), and a cytoplasmic domain (aa 1144-1257). The large extracellular domain contains six Ig-like C2 type domains and five fibronectin type III repeats. It serves as a cell recognition molecule within the nervous system that plays a critical role in multiple processes, including neuronal migration, axonal growth and fasciculation, and synaptogenesis during brain development. In the mature brain, it plays a role in the dynamics of neuronal structure and function, including synaptic plasticity. Its expression is also observed in tumor vasculature. Tumor-bearing mice that lack L1CAM exhibit reduced angiogenesis and improved vascular stabilization that leads to reduced tumor growth and metastasis. These effects have been attributed to L1CAM-induced IL-6-mediated STAT3 phosphorylation and inhibition of IL-6/JAK/STAT signaling axis that prevents endothelial cell proliferation and migration. Three isoforms of LICAM/CD171 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Magrini, A., et al. (2014). J. Clin. Invest. 124(10); 4335-4350; Maddaluno, L., et al. (2009). J. Exp. Med. Vol. 206(3); 623-635).
Physical form
Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.
Reconstitution
Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our
reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.
Storage and Stability
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.
Legal Information
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.