Skip to Content
Merck
All Photos(14)

Key Documents

ZRB2159

Sigma-Aldrich

Anti-β-Arrestin 1 Antibody, clone 1B15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Arrestin beta-1;Non-visual arresrtin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B15, recombinant monoclonal

description

1B15 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 47.07 kDa
observed mol wt ~50 kDa

purified by

using Protein A

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ARRB1(408)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B15 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects β-Arrestin 1. It targets an epitope within 22 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal region of Human β-Arrestin 1.

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected β-Arrestin 1 in MCF-7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected β Arrestin 1 in Human Lung, T47D, Raw264.7, and Rat Lung lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected β Arrestin 1 in human kidney tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected β Arrestin 1 in MCF-7 cells.

Affinity Binding Assay Analysis: A representative lot of this antibody bound beta Arrestin 1 with a KD of 2.7 x 10^-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Beta-Arrestin-1 (UniProt:P49407; also known as Arrestin beta-1, Non-visual arresrtin-2) is encoded by the ARRB1 (also known as ARR1) gene (Gene ID: 408) in human. Arrestins are a superfamily of multi-functional proteins that regulate signaling and trafficking of the majority of G-protein-coupled receptors (GPCRs), as well as sub-cellular localization and activity of many other signaling proteins. β-Arrestin can exist as a monomer or as a homodimer. Its self-association is mediated by InsP6-binding. It can also form homooligomer with β-arrestin 2 (ARRB2) that is mediated by InsP6-binding. It is detected on cytoplasm, nucleus, and membrane. The monomeric form is predominantly located in the nucleus and the oligomeric form is found in the cytoplasm. It translocates to the nucleus upon stimulation of opioid receptor 1 (OPRD1). β-arrestin 1 is reported to play a role in regulating agonist-mediated GPCR signaling by mediating both receptor desensitization and re-sensitization processes. It also targets many receptors for internalization by acting as endocytic adapters and recruiting the GPRCs to the adapter protein 2 complex 2 (AP-2) in clathrin-coated pits. However, its involvement appears to vary significantly depending on the receptor, agonist, and cell type. Internalized arrestin-receptor complexes are shown to traffic to intracellular endosomes, where they remain uncoupled from G-proteins. Two different modes of arrestin-mediated internalization have been reported. Class A receptors, such as adrenoceptor β2 and opioid receptor 1, dissociate from β-arrestin at or near the plasma membrane and undergo rapid recycling. However, class B receptors, such as arginine-vasopr+D5essin receptor 2 and neurotensin receptor 1, internalize as a complex with arrestin and traffic with it to endosomal vesicles, presumably as desensitized receptors, for extended periods of time. β-arrestin 1 is constitutively phosphorylated at serine 412 in the cytoplasm. At the plasma membrane, is rapidly dephosphorylated, a process that is required for clathrin binding and adrenoceptor β2 endocytosis but not for adrenoceptor β2 binding and desensitization. Once internalized, it is re-phosphorylated. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Deshpande, D., et al. (2008). FASEB J.22(7); 2134-2141; Beaulieu, JM., and Caron, MG. (2005). Cell. 123(5); 755-757; Lin, FT., et al. (1999). J. Biol. Chem. 274(23);15971-15974).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service