Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1940

Sigma-Aldrich

Anti-HSF1 Antibody, clone 1J20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Heat shock factor protein 1;Heat shock transcription factor 1;HSTF 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J20, recombinant monoclonal

description

1J20 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 57.26 kDa
observed mol wt ~62 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HSF1(3297)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J20 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Heat shock factor 1 (HSF1). It targets an epitope within 21 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human Heat shock factor 1 (HSF1).

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HSF1 in MCF-7 cell lysate.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HSF1 in HeLa cells.

Affinity Binding Assay: A representative lot of this antibody bound HSF1 peptide with a KD of 7.7 x 10-6 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HSF1 in human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HSF1 in MCF-7 cell lysate.

Target description

Heat shock factor protein 1(UniProt: Q00613; also known as HSF 1, Heat shock transcription factor 1, HSTF 1) is encoded by the HSF1 (also known as HSTF1) gene (Gene ID: 3297) in human. HSF1 is a stress-inducible transcription factor that plays a key role in the transcriptional activation of the heat shock response (HSR), leading to the expression of a large class of molecular chaperones heat shock proteins (HSPs). In unstressed cells, it is present in cytoplasm in its monomeric, inactive form and homotrimerizes in response to stress and translocates to the nucleus where it binds to heat shock element (HSE) sequences in promoter of heat shock protein genes and acquires transcriptional ability. In unstressed cells, its spontaneous homotrimerization is inhibited by intramolecular interactions between the hydrophobic repeat HR-A/B and HR-C regions. HSF1 also has several other functions independently of its transcriptional activity. It is reported to be involved in the repression of Ras-induced transcriptional activation of the c-Fos gene in heat-stressed cells. It is also involved in stress-induced cancer cell proliferation in a IER5-dependent manner. In unstressed cells, it remains phosphorylated on multiple serine and threonine residues. These phosphorylations are constitutive and are implicated in the repression of HSF1 activity. Its phosphorylation on serine 121 by MAPKAP kinase inhibits HSF1 homotrimerization, DNA-binding and transactivation activities. Phosphorylation on serine 303 by GSK3-β and on serine 307 by MAPK3 within the regulatory domain represses HSF1 transcriptional activity and increases its nuclear export in a CRM1 manner. Phosphorylation on serine 230 by CAM kinase 2 and on threonine 142 by casein kinase 2 is reported to enhance its transactivation activity upon heat shock. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wang, XZ., et al. (2006). J. Biol. Chem. 281(2); 782-791; Soncin, F., et al. (2003). Biochem. Biophys. Res. Comm. 303(2); 700-706; Holmberg, CI., et al. (2001). EMBO. J. 20(14); 3800-3810; Xavier, IJ., et al. (2000). J. Biol. Chem. 275(37); 29147-29152).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service