Skip to Content
Merck
All Photos(8)

Key Documents

ZRB1509

Sigma-Aldrich

Anti-V5 Tag Antibody, clone 1K20-L2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-Adiponectin Antibody, clone 1D19 ZooMAb® Rabbit Monoclonal, Non-structural protein V tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1K20-L2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 23.94 kDa

purified by

using Protein A

species reactivity

all

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1K20-L2 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects proteins with V5 tag. It targets an epitope within 14 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the internal region of V5 protein from the Parainfluenza virus 5.

Application

Quality Control Testing

Evaluated by Western Blotting with His/V5-tagged recombinant Tau-441 protein.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected His/V5-tagged recombinant Human Tau-441 protein.

Tested Applications

ELISA Analysis: A serial of dilutions from a representative lot detected V5-tagged recombinant Human Tau-441 protein.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HEK293 cells transfected with a V5 expression plasmid.

Peptide Inhibition Assay:: Target band detection in V5-tagged recombinant Human Tau-441 protein was prevented by preblocking of a representative lot with V5 tag peptide, but not the corresponding non-V5 tag control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Non-structural protein V (UniProt: P11207; also known as V5 tag) is encoded by the P/V gene in Parainfluenza virus 5. Epitope tags are genetically engineered short protein sequences that are serve as universal epitopes and can be easily detected with antibodies without affecting the native structure or function of proteins. The V5 tag is a sequence of 14 amino acids (GKPIPNPLLGLDST) derived from the internal region of non-structural protein V of parainfluenza virus 5. It can be used to detect expression of recombinant proteins in bacteria, insects, and mammalian cell systems. It is useful in detecting recombinant proteins that contain the V5 epitope. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service